    Sort by:
  • Search Within Results
(-)-Arctiin
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   Lignan derivative with antitumor activities. Down-regulates anti-apoptotic protein Bcl-XL. Potent inhibitor of HIV type-I integrase and DNA topoisomer...