    Sort by:
  • Search Within Results
AZ10417808
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   AZ10417808, non-peptidic caspase-3 inhibitor, Purity: ≥98% by HPLC, CAS Number: 331645-84-2, Molecular Formula: C₁₈H₁₃Cl₂N₅O₄, Molecular Weight: 434.23, Storage : -20 Degree C, Synonyms: 2-(3, 4-dichloroanilino)-6-nitro-4-oxo-N-prop-2-enyl-1H-quinazoline-8-carboxamide, 1mg