    Sort by:
  • Search Within Results
UNC-669
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   UNC-669 is a small molecule nicotinamide derivative that acts asan antagonist of the MBT(Malignant Brain Tumor) domain, a family of methyl-lysine bind...