    Sort by:
  • Search Within Results
Panepoxydone
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   Antibiotic. Inhibitor of NF-κB activation by preventing the phosphorylation of IκB protein and thereby interrupts the signalling pathway. Strongly inh...