You Searched For: Compound 56
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (89147-626)
Supplier: Enzo Life Sciences
Description: Potent and specific inhibitor of the tyrosine kinase activity of the epidermal growth factor receptor (EGFR) (IC50=6pM).