    Sort by:
  • Search Within Results
TMP269
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   TMP269 is a potent, selective and cell-permeable class IIa HDAC inhibitor (IC₅₀ values of of 126 nM, 80 nM, 36 nM and 19 nM for HDAC4, HDAC5, HDAC7 an...