You Searched For: 8-Bromoadenosine 3',5'-cyclic Monophosphothioate, Rp-Isomer sodium salt
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Enzo Life Sciences
Description: Inhibitor of cAMP-dependent protein kinase (PKA), preferentially binding to type I PKA. Lipophilic analog of Rp-cAMPS. Resistant to mammalian cyclic n...