    Sort by:
  • Search Within Results
1-Methyl-L-tryptophan
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (89142-916)
Supplier:  Enzo Life Sciences
Description:   Competitive inhibitor of indoleamine 2,3-dioxygenase (IDO).