    Sort by:
  • Search Within Results
Toceranib
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Toceranib is a potent, cell-permeable, ATP-competitive multi-targeted receptor tyrosine kinase inhibitor that is structurally related to Sunitinib (Ca...