    Sort by:
  • Search Within Results
MLN9708
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Selleck Chemicals
Description:   MLN9708 gets hydrolyzed to its biologically active form MLN2238, on exposure to aqueous solutions or plasma