    Sort by:
  • Search Within Results
Timentin
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Timentin is a mixture (15:1) of Ticarcillin Disodium Salt/Potassium Clavulanate. It acts as a β-Lactamase inhibitor. Highly effective combination agai...