    Sort by:
  • Search Within Results
Dabrafenib
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   Dabrafenib is a mutant BRAFV600 specific inhibitor with IC50 of 0.8 nM in cell-free assays, with 4- and 6-fold less potency agains...