You Searched For: A-967079
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (76203-484)
Supplier: Enzo Life Sciences
Description: Potent and selective TRPA1 channel blocker. Exhibits greater than 1000-fold selectivity for TRPA1 over other TRP channels. Exhibits analgesic actions.