    Sort by:
  • Search Within Results
CP-673451
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   CP-673451 is a potent inhibitor of PDGFR with IC50 value of 10 nM and 1 nM for PDGFR-α and PDGFR-β, respectively.