    Sort by:
  • Search Within Results
Suvorexant
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Suvorexant is a dual orexin receptor antagonist that blocks both OX1R and OX2R (Kis = 1.2 and 0.60 nM, respectively). It reduces locomotor activity an...