    Sort by:
  • Search Within Results
MK-8931
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   MK-8931 is a potent BACE1 inhibitor. MK-8931 binds significantly to β-secretase.