Would you like to log in again?
LigaTrap® Human IgA Purification Column, 1 mL, ImmunoReagents
Supplier: Immunoreagents
LigaTrap®
LT-146-1X1ML
LigaTrap® has developed an easier way for antibody purification in affinity purified pre-packaged columns.
  • Suitable for research and process development
LigaTrap® has developed a novel, patented series of affinity ligands specific for the purification of monoclonal and polyclonal immunoglobulins from various species. These prepacked columns are capable of processing and purifying antibodies from several matrices, including cell culture supernatant, ascites fluid, and serum.
ORDER
Expand All / Collapse All
Description Capacity Supplier No. VWR Catalog Number Unit Price Quantity
LigaTrap® Human IgA Purification Column 1×1 mL LT-146-1X1ML 76211-146 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Binding Capacity 15 mg/mL
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty