LigaTrap® Mouse IgM Purification Column, 3×1 mL, ImmunoReagents
LT-145-3X1ML
76211-144EA 477.7 USD
76211-144
LigaTrap® Mouse IgM Purification Column, 3×1 mL, ImmunoReagents
LigaTrap® has developed an easier way for antibody purification in affinity purified pre-packaged columns.
  • Suitable for research and process development
LigaTrap® has developed a novel, patented series of affinity ligands specific for the purification of monoclonal and polyclonal immunoglobulins from various species. These prepacked columns are capable of processing and purifying antibodies from several matrices, including cell culture supernatant, ascites fluid, and serum.