Somatostatin- 25, CHI Scientific Inc.
5-01949
76217-172EA 90.84 USD
76217-172
Somatostatin- 25, CHI Scientific Inc.
Ser-Asn-Pro-Ala-Met-Ala-Pro-Arg-Glu-Arg-Lys-Ala-Gly-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Thr-Ser-Cys(Disulfide bridge: Cys14-Cys25)<br>C<sub>1</sub><sub>2</sub><sub>7</sub>H<sub>1</sub><sub>9</sub><sub>1</sub>N<sub>3</sub><sub>7</sub>O<sub>3</sub><sub>4</sub>S<sub>3</sub><br>M.W.2876.36