Scyliorhinin I, Scy I; Shark Substance P Related Peptide, CHI Scientific Inc.
5-01912
76217-098EA 119.23 USD
76217-098
Scyliorhinin I, Scy I; Shark Substance P Related Peptide, CHI Scientific Inc.
Ala-Lys-Phe-Asp-Lys-Phe-Tyr-Gly-Leu-Met<br>C<sub>5</sub><sub>9</sub>H<sub>8</sub><sub>6</sub>N<sub>1</sub><sub>2</sub>O<sub>1</sub><sub>4</sub>S<br>M.W.1219.48