RGD-4C, CHI Scientific Inc.
5-01881
76217-052EA 102.19 USD
76217-052
RGD-4C, CHI Scientific Inc.
Ala-Cys-Asp-Cys-Arg-Gly-Asp-Cys-Phe-Cys-Gly (Disulfide bridge: Cys2-Cys10 . Cys4-Cys8)<br>C<sub>4</sub><sub>2</sub>H<sub>6</sub><sub>0</sub>N<sub>1</sub><sub>4</sub>O<sub>1</sub><sub>6</sub>S<sub>4</sub><br>M.W.1145.29