RF-amide, Chicken, CHI Scientific Inc.
5-01876
76217-042EA 80.9 USD
76217-042
RF-amide, Chicken, CHI Scientific Inc.
Leu-Pro-Leu-Arg-Phe-NH2<br>C<sub>3</sub><sub>2</sub>H<sub>5</sub><sub>3</sub>N<sub>9</sub>O<sub>5</sub><br>M.W.643.84