Protein Kinase G Substrate; G-Subtide, CHI Scientific Inc.
5-01842
76216-142EA 119.23 USD
76216-142
Protein Kinase G Substrate; G-Subtide, CHI Scientific Inc.
Gln-Lys-Arg-Pro-Arg-Arg-Lys-Asp-Thr-Pro<br>C<sub>5</sub><sub>3</sub>H<sub>9</sub><sub>6</sub>N<sub>2</sub><sub>2</sub>O<sub>1</sub><sub>5</sub><br>M.W.1280.74