Prosaptide 769P, CHI Scientific Inc.
5-01831
76216-120EA 79.48 USD
76216-120
Prosaptide 769P, CHI Scientific Inc.
Cys-D-Ala-Phe-Leu-Val-Lys-Glu-Val-Thr-Lys-Leu-Ile-Asp-Asn-Asn-Lys-Thr-Glu-Lys-Glu-Ile-Leu<br>C<sub>1</sub><sub>1</sub><sub>4</sub>H<sub>1</sub><sub>9</sub><sub>4</sub>N<sub>2</sub><sub>8</sub>O<sub>3</sub><sub>5</sub>S<br>M.W.2549.04