Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

proPT28, CHI Scientific Inc.

Supplier: CHI Scientific
5-01830
His-Val-Phe-Leu-Ala-Pro-Gln-Gln-Ala-Arg-Ser-Leu-Leu-Gln-Arg-Val-Arg-Arg-Ala-Asn-Thr-Phe-Leu-Glu-Glu-Val-Arg-Lys<br>C<sub>1</sub><sub>4</sub><sub>9</sub>H<sub>2</sub><sub>4</sub><sub>8</sub>N<sub>5</sub><sub>0</sub>O<sub>3</sub><sub>9</sub><br>M.W.3363.95



 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 mg 5-01830 76216-118 Each (1mg) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name proPT28
Sequence His-Val-Phe-Leu-Ala-Pro-Gln-Gln-Ala-Arg-Ser-Leu-Leu-Gln-Arg-Val-Arg-Arg-Ala-Asn-Thr-Phe-Leu-Glu-Glu-Val-Arg-Lys<br>C<sub>1</sub><sub>4</sub><sub>9</sub>H<sub>2</sub><sub>4</sub><sub>8</sub>N<sub>5</sub><sub>0</sub>O<sub>3</sub><sub>9</sub><br>M.W.3363.95
Protease-free Yes
Animal-Free Yes
Purity >95%
Endotoxin Level low
Endotoxin Content <0.1 ng/μg
Reconstitution Instructions As indicated on manual
Storage Conditions Store at -20° C prior to opening
Shipping Temperature Room Temp
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping