proPT18, CHI Scientific Inc.
5-01829
76216-116EA 70.97 USD
76216-116
proPT18, CHI Scientific Inc.
His-Val-Phe-Leu-Ala-Pro-Gln-Gln-Ala-Arg-Ser-Leu-Leu-Gln-Arg-Val-Arg-Arg<br>C<sub>9</sub><sub>6</sub>H<sub>1</sub><sub>6</sub><sub>3</sub>N<sub>3</sub><sub>5</sub>O<sub>2</sub><sub>3</sub><br>M.W.2175.59