[Pro34]-Neuropeptide Y, human, rat; [Pro34]-NPY, human, rat, CHI Scientific Inc.
5-00277
76217-414EA 193.03 USD
76217-414
[Pro34]-Neuropeptide Y, human, rat; [Pro34]-NPY, human, rat, CHI Scientific Inc.
Tyr-Pro-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Met-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Pro-Arg-Tyr- NH2<br>C<sub>1</sub><sub>8</sub><sub>9</sub>H<sub>2</sub><sub>8</sub><sub>5</sub>N<sub>5</sub><sub>5</sub>O<sub>5</sub><sub>7</sub>S<sub>1</sub><br>M.W.4271.8