Prednisone, tablets

Danger

1559505
101096-486EA 475.38 USD
101096-486
Prednisone, tablets
Prednisone
Formula: C₂₁H₂₆O₅
MW: 358.43 g/mol
MDL Number: MFCD00003608
CAS Number: 53-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR