Anti-IgG F(c) Human Polyclonal Antibody

Supplier: Immunoreagents
IR2370
10151-026EA 223.39 USD
10151-026
Anti-IgG F(c) Human Polyclonal Antibody
Antibodies
  • Origin: US

Type:
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR