Anti-IgG Goat Antibody

Supplier: Immunoreagents
GT-003-Z
10812-664EA 139.73 USD
10812-664
Anti-IgG Goat Antibody
Antibodies
Type:
Antigen: IgG
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR