Anti-IgG Donkey Antibody (HRP (Horseradish Peroxidase))

DASHRP-050
10812-634EA 765.3 USD
10812-634
Anti-IgG Donkey Antibody (HRP (Horseradish Peroxidase))
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Donkey
Isotype:
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR