You Searched For: Tool Sets
Tool sets and multi-function instruments are economical alternatives to buying individual equipment pieces. Including commonly used hand tools to achieve maintenance or adjustment needs, kits include carrying cases or boxes for the best organized storage and effortless transporting between work sites. The well equipped assortments offered can be matched to any specific environment and manual work. In logical, high-quality combinations, sets use tools that are easy to use for all skill levels.
Tool sets and multi-function instruments are economical alternatives to buying individual equipment pieces. Including commonly used hand tools to achieve maintenance or adjustment needs, kits include carrying cases or boxes for the best organized storage and effortless transporting between work sites. The well equipped assortments offered can be matched to any specific environment and manual work. In logical, high-quality combinations, sets use tools that are easy to use for all skill levels.
424  results were found
SearchResultCount:"424"
Sort Results
List View Easy View
Rate These Search Results
Supplier: VWR International
Description: Ensure the security of sanitary connections with preset torque output.
Catalog Number: (470016-710)
Supplier: Avantor
Description: Kit of small assorted tools.
Catalog Number: (470004-558)
Supplier: Avantor
Description: The eight-piece set contains one each : 16 oz. carpenter hammer; 6.5" needle nose pliers; 6" slip joint pliers; 6" adjustable wrench; 10' power tape m...
Catalog Number: (470020-436)
Supplier: Avantor
Description: Nails
Catalog Number: (97013-702)
Supplier: PERKINELMER U.S. LLC
Description: Made of soft alloy copper, these cleaning wires are specially deburred to prevent scratching of the orifices and internal components of the nebulizers...
Supplier: Thermo Scientific
Description: This tool kit contains wrench sets, flashlight, brushes, mini-drill set and other tools for maintaining GC system performance.
Supplier: Thermo Scientific
Description: Support GC applications.
Catalog Number: (10138-790)
Supplier: PERKINELMER U.S. LLC
Description: Shipped in a plastic tool box for convenient storage.
Catalog Number: (89219-962)
Supplier: MicroSolv Technology Corp
Description: Tool kit for high pressure HPLC columns.
Catalog Number: (10753-252)
Supplier: Micronova
Description: A custom tool specifically designed to hold swabs for environmental monitoring and sampling. Use with SSU series handles for extension in large tanks ...
Small Business Enterprise
Catalog Number: (10204-914)
Supplier: Bel-Art Products, a Part of SP
Description: With five hole sizes ranging from 3.5 mm to 14.5 mm, this holder accommodates various thermometers including liquid in glass, electronic with probe, a...
Catalog Number: (95044-450)
Supplier: Cytiva
Description: Separation tool.
Product available on GSA Advantage®
Catalog Number: (10185-366)
Supplier: Sklar
Description: D'errico Enlarging Burrs are industry standard and constructed of stainless steel.
Catalog Number: (10194-938)
Supplier: Sklar
Description: This is a dermal instrument commonly used for drilling a nailbed to relieve blood or pressure from underneath the nail.
Catalog Number: (10195-800)
Supplier: Sklar
Description: This is cannulated with Jacobs Chuck for twist drills and Kirschner wires.
Catalog Number: (10195-792)
Supplier: Sklar
Description: This drill set is constructed of stainless steel.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 424
no targeter for Bottom