You Searched For: Screwdrivers
Manual or electric-powered screwdrivers feature tips that are shaped and sized to fit common grooves or slots. Comfortable handles making the torque generated from turning successfully insert the threaded devices. Economical kits include interchangeable bits for necessary modifications to accommodate any screw. Handles may be color coded to guarantee the correct size is selected, as damage to the screw is possible if incorrect tool is used.
Manual or electric-powered screwdrivers feature tips that are shaped and sized to fit common grooves or slots. Comfortable handles making the torque generated from turning successfully insert the threaded devices. Economical kits include interchangeable bits for necessary modifications to accommodate any screw. Handles may be color coded to guarantee the correct size is selected, as damage to the screw is possible if incorrect tool is used.
132  results were found
SearchResultCount:"132"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (76191-480)
Supplier: Sklar
Description: All Sklar products are industry standard.
Supplier: WIHA TOOLS TE
Description: These screwdrivers are designed with precision handle made of cadmium free, high quality plastic. Blade is made from high alloy chrome-vanadium molybd...
Catalog Number: (300030-482)
Supplier: Safety & Industrial Supplies
Description: Handle marked for easy identification of screwdriver size, type, and length.
Catalog Number: (82030-534)
Supplier: Thermo Fisher Scientific
Description: Quickly and easily cap and decap Nunc Cryobank tubes using the manual Nunc™ Screwdriver for Cryobank and Bank-It™ Systems
Supplier: Cooper Tools
Description: These screwdrivers feature hard shockproof plastic handle core for durability, while the outer coating is soft rubber for comfort and slip resistance.
Catalog Number: (76254-192)
Supplier: HISCO, INC TE
Description: This is designed ergonomically and dual component handle for increased comfort, control and safety.
Catalog Number: (300002-508)
Supplier: Safety & Industrial Supplies
Description: Accurate fit and more torque without slippage.
Catalog Number: (76248-196)
Supplier: TESTEQUITY LLC TE
Description: A Proto® power socket bit holder for 1/4" drives. Not for use in impact applications. Black oxide finish prevents corrosion.
Supplier: Aven Tools
Description: Specifically designed for electronics
Supplier: Electron Microscopy Sciences
Description: The hexagonal swivel heads prevent rolling and knurled handles for a firm grip. Color identifier bands allow for easy selection and set screws hold th...
Minority or Woman-Owned Business Enterprise
Supplier: Electron Microscopy Sciences
Description: A very handy set of Phillips screwdrivers which feature one of the smallest sizes available (1.20 mm). A knurled body with hexagonal heads and tempere...
Minority or Woman-Owned Business Enterprise
Supplier: Safety & Industrial Supplies
Description: BIT INSERT SLOT 5/16HEX
Catalog Number: (76243-948)
Supplier: Cooper Tools
Description: These screwdrivers handle more jobs with fewer tools, saves bench space and lightens service kit.
Supplier: WIHA TOOLS TE
Description: The ideal multi-purpose bits for all around quality and performance.
Catalog Number: (76249-830)
Supplier: WIHA TOOLS TE
Description: Chrome-vanadium-molybdenum S2 modified tool steel.
Supplier: Safety & Industrial Supplies
Description: Precision machined tip for proper screw engagement.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 132
no targeter for Bottom