QuickDetect™ Catecholamine (CA) (Mouse) ELISA Kit, BioVision

Supplier: BioVision
QuickDetect™
AB287795-96TEST
76211-184EA 1049.53 USD
76211-184
QuickDetect™ Catecholamine (CA) (Mouse) ELISA Kit, BioVision
Assays ELISAs
A rapid ELISA kit to quantify Catecholamine in Mouse serum, plasma, urine, cell culture samples

  • Easy, convenient and time-saving method to measure the level of Catecholamine in mouse Serum, plasma, urine, cell culture samples, biological fluid.
  • Detection Range: 5 - 320 pg/ml
  • Sensitivity: < 1.2 pg/ml
  • Cross Reactivity: No significant cross-reactivity or interference between this analyte and its analogues was observed.

Catecholamine (CA) is a monoamine, an organic compound that has a catechol and a side-chain amine. Catecholamines are derived from the amino acid tyrosine, which is derived from dietary sources as well as synthesis from phenylalanine. Two catecholamines, norepinephrine and dopamine, act as neuromodulators in the central nervous system and as hormones in the blood circulation. The catecholamine norepinephrine is a neuromodulator of the peripheral sympathetic nervous system but is also present in the blood. It causes general physiological changes that prepare the body for physical activity. Some typical effects are increases in heart rate, blood pressure, blood glucose levels, and a general reaction of the sympathetic nervous system. BioVision’s Catecholamine ELISA kit is a sandwich ELISA assay for the quantitative measurement of CA in mouse serum, plasma and cell culture supernatants in 90 minutes. The density of color is proportional to the amount of Catecholamine captured from the samples.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR