SP Wilmad-LabGlass Vacuum Adapters with 105° Bend, Style B, Standard Taper Joints, SP Industries
Supplier: SP Industries
ML-390-702
14210-522EA 149.46 USD
14210-522
SP Wilmad-LabGlass Vacuum Adapters with 105° Bend, Style B, Standard Taper Joints, SP Industries
Adapters Adapters, Conical Ground Joints
These borosilicate glass vacuum-type adapters feature Standard Taper joints (top outer and bottom inner) and a sealed-in inner delivery tube on the bottom joint which ends in a slanted drip tip.
Designed with a side hose connector for connection to a vacuum source.