SP Wilmad-LabGlass Reaction Flask Heating Mantles, SP Industries
Supplier: SP Industries
LG-8879-104 LG-8879-108 LG-8879-102 LG-8879-106 LG-8879-100 LG-8879-110
14200-928EA 680.55 USD
14200-928 14200-794 14200-860 14200-896 14200-856 14200-858
SP Wilmad-LabGlass Reaction Flask Heating Mantles, SP Industries
These all-glass constructed heating mantles are for use with LG-8010 and LG-8011 reaction flasks. They provide even heat distribution and prevent scorching of clear materials.
  • All heat input is in one electrical circuit to simplify temperature control
  • Depth given is the interior measurement