DuPont™ Tychem® 6000 Sleeves
Supplier: DuPont
TFPS32TGY0000500
76205-422CS 138.58 USD
76205-422
DuPont™ Tychem® 6000 Sleeves
Disposable Garments
Tychem® 6000 garments are made of a proprietary barrier film laminated to a Tyvek® substrate.
  • Stiched and over-taped seams
  • Strong, durable, and lightweight
The fabric provides at least 30 minutes of protection against over 181 challenge chemicals including chemical warfare agents and toxic industrial chemicals. Tychem® 6000 garments are strong, durable and lightweight, and they are available in both high-visibility orange and low-visibility gray fabrics, making them a preferred choice for law enforcement, emergency medical services (EMS) technicians and military personnel. Sleeves feature a 8 1/2" bicep opening.