Willett-Martel Scalp Flap Forceps, OR Grade, Sklar®
Supplier: Sklar
17-1059
76192-184EA 134.42 USD
76192-184
Willett-Martel Scalp Flap Forceps, OR Grade, Sklar®
Forceps
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar's® Willet-Martel (also Martel or De Martel) scalp flap forceps are used in the obstetrics field to ease the deliver of a newborn by grasping its scalp with the smooth flattened blades.