    Sort by:
  • Search Within Results
(R)-(-)-2-Aminooctane
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   (R)-(-)-1-Methylheptylamine (2R)-Octylamine. Grade: ChiPros\(tm 99+%, ee 99+. Melting Point C<-20*. Boiling Point C: 163-165*. C8H19N. 34566-05-7. Trademark of BASF AG. FLAMMABLE TOXIC CORROSIVE AIR SENSITIVE