Would you like to log in again?
Penicillin : Streptomycin : L-glutamine 25k/25k
Supplier: LONZA WALKERSVILLE INC
17-718R
Penicillin-Streptomycin-L-glutamine is a broad spectrum bacteriostatic and bactericidal, with activity
against gram negative and gram positive organisms. It also contains the essential amino acid L-glutamine which is necessary for cell culture growth. Upon thawing, activity decreases rapidly.
  • Working concentration: 10 mL per liter
  • Store penicillin-streptomycin mixtures at –4 to 14 °F (–20 to –10 °C )
The working concentration of Penicillin-streptomycin-L glutamine
25,000 units/ml potassium penicillin
25,000 mcg/ml streptomycin sulfate
29.2mg L-glutamine per mL
Caution: This product is for research use only. Not recommended for human, veterinary use, or for use in clinical or in vitro diagnostic procedures.
ORDER
Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
25 x 4.5 mL 17-718R 12001-352 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty