Carbenicillin solution 50 mg/ml,

C2125
CA100216-946
Carbenicillin solution 50 mg/ml,
Carbenicillin

Carbenicillin is a semi-synthetic ampicillin analog, and like ampicillin is a beta-lactam antibiotic. Carbenicillin is a member of the carboxypenicillin subgroup of the penicillins.


It is effective against Gram-negative organisms including H. influenzae and E. coli. Carbenicillin is also used in transformed Agrobacter species. Carbenicillin was the first penicillin with significant activity against Pseudomonas species.

Formula: C₁₇H₁₈N₂O₆S
CAS Number: 4697-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR