Avantor Single-Use C-Flex Adapters and Extensions, Avantor Fluid Handling
5000214 5000160 5000224 5000218 5000157 5000220 5000221 5000217 5000211 5000213 5000223 5000172 5000215 5000219 5000166 5000222 5000216 5000163 5000212 5000169
76124-604CS 437.14 USD
76124-604 76124-626 76124-636 76124-614 76124-628 76124-606 76124-616 76124-638 76124-608 76124-618 76124-640 76124-642 76124-620 76124-630 76124-622 76124-610 76124-632 76124-624 76124-612 76124-634
Avantor Single-Use C-Flex Adapters and Extensions, Avantor Fluid Handling
Sterile C-Flex Adapters and Extensions are designed to facilitate and expedite the on-site construction of single-use fluid transfer assemblies.
  • Manufactured in ISO Class 7 Cleanroom
  • Supplied Gamma Irradiated to Sterility Assurance Level (SAL) 10-6
Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, and production and cleanroom applications.
Assembled in an ISO Class 7 cleanroom and manufactured from USP Class VI materials. Tubing assemblies are offered with custom size and configuration capabilities. Tubing is weldable/sealable. All product contact materials are animal-free or processed in accordance to EMEA/410/01.
The assemblies are able to withstand temperatures from –20 to 65 °C (– 4 to 149 °F) without losing integrity.
When used with sterile Avantor Bottle or Flask Assemblies, custom possibilities are endless.
Packaging: All assemblies are individually double-bagged with product label located on inner bag.