    Sort by:
  • Search Within Results
You Searched for: "silicone-oil-12-500-cst-25c-polydimethylsiloxane"
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Brookfield
Description:   Calibrated at 25C (77F); used for checking calibration of most Viscometers/Rheometers. Silicone fluids have an accuracy of +/-1% and are NIST traceable. Supplied in one pint container with certificate of calibration.