pFN23A HaloTag CMVd2 Flexi Vector, 20 µg, Promega
Supplier: Promega
G2861
PAG2861EA 278.25 USD
PAG2861
pFN23A HaloTag CMVd2 Flexi Vector, 20 µg, Promega
Vectors, Plasmids and Libraries
Vectors designed for expression of N-terminal-tagged HaloTag fusion proteins in mammalian cells. Once expressed, use the HaloTag fusion protein for cell imaging of protein localization or trafficking in conjunction with the fluorescent HaloTag Ligands.
  • Appends HaloTag(R) to N-Terminus of Protein Fusion Partner
  • Enables constitutive protein expression in mammalian cells
  • Includes a modified CMVd2 immediate-early enhancer/promoter
  • Suited to stable or transient gene expression
These vectors are designed for expression of N-terminal-tagged HaloTag fusion proteins in mammalian cells. Once expressed, the HaloTag fusion protein may be used for cell imaging of protein localization or trafficking in conjunction with the fluorescent HaloTag Ligands. In addition, the HaloTag fusion protein can be purified or pulled down as a complex with its protein partners. We offer two types of HaloTag fusion vectors to accommodate your cloning preferences: pHT Vector Series: Simple Multiple Cloning Site (MCS) plasmids for traditional cloning. pF Vector Series: Flexi Vector Cloning System---a directional cloning method for protein-coding sequences. It is based on two rare-cutting restriction enzymes, SgfI and PmeI, and provides a rapid, efficient and high-fidelity way to transfer protein-coding regions between a variety of Flexi Vectors without the need to resequence. Note: Flexi Vectors contain the lethal barnase gene to reduce background colonies without inserts during the subcloning procedure. Using the Flexi Vector Cloning System replaces the barnase gene with your insert. These vectors, as purchased, cannot be cultured in normal laboratory strains of E. coli without an insert. Find your gene, precloned, and experimentally validated at: www.promega.com/products/pm/halotag-technology/kazusa-collection