ABT-199 ≥99% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
2-((1H-Pyrrolo[2,3-b]pyridin-5-yl)oxy)-4-(4-((4'-chloro-5,5-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)-N-((3-nitro-4-(((tetrahydro-2H-pyran-4-yl)methyl)amino)phenyl)sulfonyl)benzamide
, 2-((1H-Pyrrolo[2,3-b]pyridin-5-yl)oxy)-4-(4-((4'-chloro-5,5-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)-N-((3-nitro-4-(((tetrahydro-2H-pyran-4-yl)methyl)amino)phenyl)sulphonyl)benzamide
, Venetoclax
Venetoclax is a Bcl-2-selective inhibitor with Ki of <0.01 nM, >4800-fold more selective versus Bcl-xL and Bcl-w, and no activity to Mcl-1
- Bcl-2 selective inhibitor
Yellow solid.
Soluble in DMSO.
Formula:
C₄₅H₅₀ClN₇O₇S MW: 868,44 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD23160052 CAS Number: 1257044-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...