Sonidegib free base ≥99% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
LDE225
, N-(6-(cis-2,6-Dimethylmorpholino)pyridin-3-yl)-2-methyl-4'-(trifluoromethoxy)-[1,1'-biphenyl]-3-carboxamide
CHM238-0050
CHM238-0500
CHM238-0025
CHM238-0010
CHM238-0100
CHM238-0200
76203-294EA
186.32
USD
76203-294
76203-404
76203-402
76203-400
76203-298
76203-296
Sonidegib free base ≥99% (by HPLC)
Sonidegib
Sonidegib, also known as LDE-225, is a potent and selective smoothened (Smo) antagonist.
- Smoothened antagonist
White solid.
Soluble in DMSO.
Formula:
C₂₆H₂₆F₃N₃O₃ MW: 485,5 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD16038928 CAS Number: 956697-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...