SureVector Library Cloning Kit, Agilent Technologies
Supplier: Agilent Technologies
G7556A G7556A
76193-686EA 1605.67 USD
CA76193-686 76193-686
SureVector Library Cloning Kit, Agilent Technologies
Vectors, Plasmids and Libraries
SureVector library cloning kit contains all items to clone a CRISPR library from a pooled of oligos.
  • Plasmid libraries generated optimized to work in mammalian cells with expression driven by a human U6 promoter
  • SureVector cloning technology adapted to insert a custom library into the lentiviral plasmid pSGLenti
The library cloning kit uses Agilent’s SureVector chemistry to allow the rapid generation of plasmid libraries from an amplified oligo pool. Included in the kit are Agilent's linearized pSGLenti vector backbone, competent cells, media, and all buffers and reagents required to clone Agilent's pooled ready-to-clone or ready-to-amplify libraries.
pSGLenti is directly compatible with Agilent's ready-to-clone libraries while alternative backbones, supplied separately, can also be used to clone fully custom, ready-to-amplify libraries. Each kit contains sufficient amounts of the above materials to generate four plasmid libraries of 60000 guides.
Caution: For research use only. Not for use in diagnostic procedures.