Dabrafenib methanesulfonate salt ≥99% (by HPLC)
Synonyms:
N-(3-(5-(2-Aminopyrimidin-4-yl)-2-(tert-butyl)thiazol-4-yl)-2-fluorophenyl)-2,6-difluorobenzenesulphonamide methanesulphonate
, GSK-2118436 mesylate
, N-(3-(5-(2-Aminopyrimidin-4-yl)-2-(tert-butyl)thiazol-4-yl)-2-fluorophenyl)-2,6-difluorobenzenesulfonamide methanesulfonate
CHM220-1000
CHM220-0100
CHM220-2000
CHM220-0500
CHM220-0050
CHM220-0010
CHM220-0200
76203-174EA
1885.25
USD
76203-174
76203-162
76203-172
76203-170
76203-168
76203-166
76203-176
Dabrafenib methanesulfonate salt ≥99% (by HPLC)
Dabrafenib methanesulfonate salt
Dabrafenib methanesulfonate salt is a selective BRAF V600E inhibitor. Dabrafenib methanesulfonate salt binds to Raf family kinases and inhibits their activity.
- Selective BRAF V600E inhibitor
White solid.
Soluble in DMSO.
Formula:
C₂₄H₂₄F₃N₅O₅S₃ MW: 615,67 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD20922872 CAS Number: 1195768-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...