A-967079 ≥99% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: (1E,3E)-1-(4-Fluorophenyl)-2-methyl-1-penten-3-one oxime

CHM210-1000
76203-484EA 2071.58 USD
76203-484
A-967079 ≥99% (by TLC)
A-967079

Potent and selective TRPA1 channel blocker. Exhibits greater than 1000-fold selectivity for TRPA1 over other TRP channels. Exhibits analgesic actions.


  • TRPA channel blocker


White solid.


Soluble in DMSO, DMF or ethanol.

Storage Temperature: Freezer
CAS Number: 1170613-55-4
https://us.vwr.com/stibo/web/std.lang.all/17/00/45401700.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR