Siguazodan ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-PD179-0005
76203-480EA 148.02 USD
76203-480
Siguazodan ≥98% (by HPLC)
Siguazodan

A selective phosphodiesterase PDE III inhibitor, IC50=0.7µM. Siguazodan is a potent inotropic, vasodilator and antiplatelet agent with sustained duration of action. It synergizes with PDE IV inhibitors (roliprams) in blocking antigen-induced bronchospasm in the guinea pig.


  • PDE inhibitor


White solid.


Soluble in DMSO.

Storage Temperature: Ambient
CAS Number: 115344-47-3
https://us.vwr.com/stibo/web/std.lang.all/14/13/45401413.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR